Orders LinkContactUsLinkComponent
Multidishes, non treated, Nunc™
Multidishes, non treated, Nunc™
48-brønds multiskål, 0,5 ml, Pakket enkeltvis
Katalog # 735-0225
Leverandør:   Thermo Fisher Scientific
Multidishes, non treated, Nunc™
Katalog # 735-0225
Multidishes, non treated, Nunc™
Multidishes, non treated, Nunc™
48-brønds multiskål, 0,5 ml, Pakket enkeltvis
Price
3.280,00 kr
Log in for availability
1 * 75 Items
Product Details & Documents
PS, non treated, with lid, sterile.
  • Raised well rims to lower risk of cross contamination
  • Excellent optical quality
  • Non pyrogenic
Specifications
  • Pakket:
    Pakket enkeltvis
  • Anbefalet arbejdsvolumen:
    0,5 ml
  • Beskrivelse:
    48-brønds multiskål