Multidishes, non treated, Nunc™
48-brønds multiskål, 0,5 ml, Pakket enkeltvis
Katalog # 735-0225
Leverandør: Thermo Fisher Scientific
Jump to:
Product Details & Documents
PS, non treated, with lid, sterile.
- Raised well rims to lower risk of cross contamination
- Excellent optical quality
- Non pyrogenic
Specifications
- Pakket:Pakket enkeltvis
- Anbefalet arbejdsvolumen:0,5 ml
- Beskrivelse:48-brønds multiskål