Orders LinkContactUsLinkComponent
GC ferrules, SilTite™
GC ferrules, SilTite™
For 0,25 mm ID columns
Katalog # 555-9246
Leverandør:   Thermo Fisher Scientific
GC ferrules, SilTite™
Katalog # 555-9246
GC ferrules, SilTite™
For 0,25 mm ID columns
Price
904,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
SilTite™ ferrules are designed for use with capillary column connectors.
Specifikationer
  • Beskrivelse:
    For 0,25 mm ID columns
  • Seq:
    01