Reacti-Therm™ modules
Reacti-Therm™ module with heating function, for 27 port sample concentrator, 3, Ambient +10...200 °C
Katalog # HYPETS-18824
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Specifikationer
- Antal blokke:3
- Temperaturområde:Ambient +10...200 °C
- Beskrivelse:Reacti-Therm™ module with heating function, for 27 port sample concentrator
- Seq:014
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Remote temperature probe, for Reacti-Therm™ heating and stirring modulesRemote temperature probe, for Reacti-Therm™ heating and stirring modulesHYPETS-18820Log ind for tilgængelighed1 * 1 Items - 3.220,00 kr