Reacti-Therm™ modules
Reacti-Therm™ module with heating function, for 9 port sample concentrator, 1, Ambient +10...200 °C
Catalog # HYPETS-18822
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Specifications
- Antal blokke:1
- Temperaturområde:Ambient +10...200 °C
- Beskrivelse:Reacti-Therm™ module with heating function, for 9 port sample concentrator
- Seq:012
Accessories
Product Information
- Catalog #BeskrivelseAvailabilityPrice
- HYPETS-18820Remote temperature probe, for Reacti-Therm™ heating and stirring modulesLog in for availability1 * 1 Items - 3.220,00 kr