Screw cap septum vials
7,00 ml, Flat bottom, 17×60 mm, klar, No
Catalog # HYPETS-13028
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
These vials are ideal for sample collection and small derivatisation reactions for use with automated gas (GC) and liquid chromatography (LC), and for storage of reagents and standards under complete seal with instant syringe access. They are made of autoclavable borosilicate glass.
- Flat bottom
Specifications
- Kasket inkluderet:No
- Diameter×H:17×60 mm
- Kapacitet:7,00 ml
- Farve:klar
- Seq:02
- Grundlag:Flat bottom



