PTFE/rubber laminated discs
PTFE/rubber laminated disc, 0,3 and 1,0 ml Reacti-Vial™ small reaction vials, 2 and 3,5 ml screw cap septum vials, 12 mm, 1,5 mm
HYPETS-12412
These discs are designed to provide a highly inert and unreactive seal. Constructed of white pharmaceutical rubber with 0,1 mm of PTFE coating bonded to one side, they are excellent for use as cap liners when a highly inert non reactive seal is desired.
- Autoclavable with no loss of integrity after heating above 100 °C for five hours
Discs are rigid and more difficult to puncture than PTFE/silicone discs.
- Ø:12 mm
- Til:0,3 and 1,0 ml Reacti-Vial™ small reaction vials, 2 and 3,5 ml screw cap septum vials
- Beskrivelse:PTFE/rubber laminated disc
- Seq:01
- Tykkelse:1,5 mm



