Orders LinkContactUsLinkComponent
Inlet base seals
Inlet base seals
Siltek®-treated inlet base seals, 0,8 mm ID
Katalog # 555-9221
Leverandør:   Thermo Fisher Scientific
Inlet base seals
Katalog # 555-9221
Inlet base seals
Inlet base seals
Siltek®-treated inlet base seals, 0,8 mm ID
Price
724,00 kr
Log ind for tilgængelighed
1 * 2 Items
Product Details & Documents
Ideal for analysis of active compounds such as pesticides, polychlorinated biphenyls (PCBs) and phenol.
Compatible with all Agilent GC 5890/6890/7890 split/splitless injection ports.
Specifikationer
  • Beskrivelse:
    Siltek®-treated inlet base seals, 0,8 mm ID
  • Seq:
    08