0,00 kr
Ordreindtastning
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
Orders Link
ContactUsLinkComponent
Sprøjtenåle
Needles for Reacti Vap sample concentrator
Needles for Reacti Vap sample concentrator
Stainless steel PTFE coated needles, blunt end, length: 102 mm, 19, 102 mm
HYPETS-18784
:
Thermo Fisher Scientific
Needles for Reacti Vap sample concentrator
HYPETS-18784
:
Thermo Fisher Scientific
Needles for Reacti Vap sample concentrator
Stainless steel PTFE coated needles, blunt end, length: 102 mm, 19, 102 mm
2.150,00 kr
1 * 9 Items
-
+
Måler:
19
Beskrivelse:
Stainless steel PTFE coated needles, blunt end, length: 102 mm
Længde:
102 mm
Seq:
031
Reagents for Karl Fischer titrations, Aquastar®, Supelco®
#: 1.88015.1000
: Merck