Orders LinkContactUsLinkComponent
Magnetic stirring bars, PTFE, triangular, Spinstar®
Triangular, Reacti-Vial™ small reaction vials, 0,3 and 1,0 ml
Katalog # HYPETS-16010
Leverandør:   Thermo Fisher Scientific
Magnetic stirring bars, PTFE, triangular, Spinstar®
Katalog # HYPETS-16010
Magnetic stirring bars, PTFE, triangular, Spinstar®
Triangular, Reacti-Vial™ small reaction vials, 0,3 and 1,0 ml
Price
2.090,00 kr
Log ind for tilgængelighed
1 * 6 Items
Product Details & Documents
Triangular stir bars can be used for faster reaction times and smooth mixing of small samples in conical-bottom vials of the Thermo Scientific™ Reacti-Therm™ Derivatization System. The stir bars provide simultaneous stirring of samples in multiple vials when used with a Reacti-Therm Heating/Stirring Module equipped with appropriate Reacti-Block Aluminium Blocks.
Specifikationer
  • Til:
    Reacti-Vial™ small reaction vials, 0,3 and 1,0 ml
  • Model:
    Triangular
  • Seq:
    01