Orders LinkContactUsLinkComponent
Drenchers
Drenchers
Drencher, straight, 203 mm, 3,0 mm
Catalog # SOCO376.30203S
Supplier:   Socorex Isba
Drenchers
Catalog # SOCO376.30203S
Supplier:  Socorex Isba
Drenchers
Drenchers
Drencher, straight, 203 mm, 3,0 mm
Price
318,00 kr
Log ind for tilgængelighed
1 * 2 Items
Product Details & Documents
These straight or bent drenchers are made from stainless steel. They are ideal for safe animal nose and mouth feeding.
  • Luer lock fittings
Specifikationer
  • Udv.Ø:
    3,0 mm
  • Beskrivelse:
    Drencher, straight
  • Længde:
    203 mm
  • Seq:
    08