Orders LinkContactUsLinkComponent
UHPLC filter
2,1 mm ID replacement filter cartridge, 0,2 µm
Katalog # 555-7042
Leverandør:   Thermo Fisher Scientific
UHPLC filter
Katalog # 555-7042
UHPLC filter
UHPLC filter
2,1 mm ID replacement filter cartridge, 0,2 µm
Price
2.350,00 kr
Log ind for tilgængelighed
1 * 5 Items
Product Details & Documents
Column protection for Hypersil GOLD™ 1,9 µm and Syncronis™ 1,7 µm columns without compromising performance.
  • Low volume filter cartridge design
  • Maintain peak shape
  • Minimal efficiency loss through dispersion
Specifikationer
  • Porøsitet:
    0,2 µm
  • Beskrivelse:
    2,1 mm ID replacement filter cartridge
  • Seq:
    02