Methylcyclohexan for synthesis, Sigma-Aldrich®
Catalog # 8.06147.6010
Supplier: Merck
CAS Number: 108-87-2
Jump to:
Product Details & Documents
- CAS Number:108-87-2
- Formula:C₆H₁₁CH₃
- MDL Number:MFCD00001497
- Molecular Weight:98.19 g/mol
- Merck Index:13,06074
- Boiling Pt:101 °C (1013 hPa)
- Melting Pt:–126 °C
- Density:0,7693 g/cm³ (20 °C)
- Flash Pt:–4 °C
- Storage Temperature:Stue temperatur
- UN:2296
- ADR:3,II
- :M-Clarity™ quality level = MQ200
- EINECS:203-624-3
Documents
Certificates
Search by Lot NumberMERC8.06147.1000_TDS_EN ProductPage_806147
Product specification
msds_806147
SDS
Specifications
Specification Test Results
- Solubility:0.014 g/l (20 °C)
- Melting Point:-126 °C
- Molar Mass:98.19 g/mol
- Boiling Point:100.9 °C (1013 hPa)
- Vapor Pressure:48 hPa (20 °C)
- Flash Point:-4 °C
- Explosion Limit:1.1 - 6.7 %(V)
- Density:0.77 g/cm3 (20 °C)
- Ignition Point:260 °C
- Hill Formula:C7H14
- Chemical Formula:C6H11CH3
Specifications
- Pakning:Stainless steel drum, returnable
- Pakn:10 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



