[EN]MICRO TUBES 1.5ML SKIRTED ASSEMB PRI 1 * 1.000 Items
Catalog # SARS72.703.406
Supplier: SARSTEDT
[EN]MICRO TUBES 1.5ML SKIRTED ASSEMB PRI 1 * 1.000 Items
Catalog # SARS72.703.406
Supplier: SARSTEDT
Frequently Bought Together
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® Blender Bags
Supplier: VWR Collection
VWR®, Spredere, T-formede
Supplier: VWR Collection
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
Serologiske pipetter, Stripette™
Supplier: Corning
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
VWR®, Serologiske pipetter, Standard type
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
Screw Caps for VWR® Scintillation Vials
Supplier: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR®, Overførselspipetter, Standard Line
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
Blood collecting system, S-Monovette® LH (on beads)
Supplier: SARSTEDT