Jump to:
Product Details & Documents
Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Number:143-74-8
- Formula:C₁₉H₁₄O₅S
- MDL Number:MFCD00003552
- Molecular Weight:354.38 g/mol
- Merck Index:13,07329
- Melting Pt:> 300 °C
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Stue temperatur
- EINECS:205-609-7
Documents
Certificates
Search by Lot NumberMERC1.07241.0005_TDS_EN MERC1.07241.0025_TDS_EN ProductPage_107241
Product specification
msds_107241
SDS
Specifications
Specification Test Results
- Solubility:0.77 g/l (100 °C)
- Melting Point:300 °C
- Molar Mass:354.38 g/mol
- Bulk Density:200 - 300 kg/m3
Specifications
- Pakning:Metal can
- Pakn:1 kg



