Waste caps
Waste cap, includes O-ring, nuts, ferrules, plugs and colour-sleeve adapter, B 83, 4 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with M10 x 1 thread for barbed adapter, PP
Katalog # JOURJR-S-18314
Leverandør: VICI JOUR RESEARCH
Gå til:
Product Details & Documents
These waste caps include three or four ports for ¹/₈ and ¹/₁₆" tubing. Two versions of each waste cap are available, with an extra port to accommodate either a 1/2-20 port for ¹/₄" O.Ø tubing, or a barbed hose adapter.
Bestillingsoplysninger: Barbed adapters must be ordered separately.
Specifikationer
- Gevind:B 83
- Beskrivelse:Waste cap, includes O-ring, nuts, ferrules, plugs and colour-sleeve adapter
- Materiale:PP
- Seq:04
- Stik:4 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with M10 x 1 thread for barbed adapter