Waste caps
Waste cap, includes O-ring, nuts, ferrules, plugs and colour-sleeve adapter, DIN 60, 3 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with M10 x 1 thread for barbed adapter, PP
Catalog # JOURJR-S-16013
Supplier: VICI JOUR RESEARCH
Jump to:
Product Details & Documents
These waste caps include three or four ports for ¹/₈ and ¹/₁₆" tubing. Two versions of each waste cap are available, with an extra port to accommodate either a 1/2-20 port for ¹/₄" O.Ø tubing, or a barbed hose adapter.
Ordering information: Barbed adapters must be ordered separately.
Specifications
- Gevind:DIN 60
- Beskrivelse:Waste cap, includes O-ring, nuts, ferrules, plugs and colour-sleeve adapter
- Materiale:PP
- Seq:09
- Stik:3 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with M10 x 1 thread for barbed adapter