Sulfonation flasks, with three necks
2500 ml, 45/40 NS, 29/32 NS
Katalog # WITG0.851.250
Leverandør: witeg Labortechnik
Gå til:
Product Details & Documents
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With one parallel and two angled side necks.
Specifikationer
- Kapacitet:2500 ml
- Seq:06
- Sidehalse:29/32 NS
- Centreret hals:45/40 NS