Orders LinkContactUsLinkComponent
Sulfonation flasks, with three necks
2500 ml, 45/40 NS, 29/32 NS
Katalog # WITG0.851.250
Leverandør:   witeg Labortechnik
Sulfonation flasks, with three necks
Katalog # WITG0.851.250
Leverandør:  witeg Labortechnik
Sulfonation flasks, with three necks
Sulfonation flasks, with three necks
2500 ml, 45/40 NS, 29/32 NS
Price
3.090,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With one parallel and two angled side necks.
Specifikationer
  • Kapacitet:
    2500 ml
  • Seq:
    06
  • Sidehalse:
    29/32 NS
  • Centreret hals:
    45/40 NS