Orders LinkContactUsLinkComponent
Storage bags
Storage bags
Storage bags, transparent, with ground weld., 0,05 mm, 900×900 mm
Katalog # SEMA4447
Leverandør:   SEMADENI
Storage bags
Katalog # SEMA4447
Leverandør:  SEMADENI
Storage bags
Storage bags, transparent, with ground weld., 0,05 mm, 900×900 mm
Price
1.400,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
Storage bags in transparent LDPE, with bottom weld.
They are also suitable for liquids but without side weld.
Emballering: Minimum quantity:100 pieces
Specifikationer
  • Beskrivelse:
    Storage bags, transparent, with ground weld.
  • Dimensioner:
    900×900 mm
  • Seq:
    10
  • Tykkelse:
    0,05 mm