Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for Assortment Boxes, PS

Accessories for Assortment Boxes, PS

Compartments, PS for storage boxes, yellow, 108×54 mm, 45 mm height
Katalog # SEMA0436
Leverandør:   SEMADENI
Accessories for Assortment Boxes, PS
Katalog # SEMA0436
Leverandør:  SEMADENI
Accessories for Assortment Boxes, PS
Accessories for Assortment Boxes, PS

Compartments, PS for storage boxes, yellow, 108×54 mm, 45 mm height

Price
25,10 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Compartments, PS for storage boxes, yellow, 108×54 mm, 45 mm height
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.