Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Accessories for Assortment Boxes, PS

Compartments, PS for storage boxes, red, 54×54 mm, 45 mm height
Katalog # SEMA0435
Leverandør:   SEMADENI
Accessories for Assortment Boxes, PS
Katalog # SEMA0435
Leverandør:  SEMADENI
Accessories for Assortment Boxes, PS
Accessories for Assortment Boxes, PS

Compartments, PS for storage boxes, red, 54×54 mm, 45 mm height

Price
22,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Compartments, PS for storage boxes, red, 54×54 mm, 45 mm height
  • Seq:
    02