Orders LinkContactUsLinkComponent
Jars with overlapping lid
39 mm, 15 mm
Katalog # SEMA2961
Leverandør:   SEMADENI
Jars with overlapping lid
Katalog # SEMA2961
Leverandør:  SEMADENI
Jars with overlapping lid
Jars with overlapping lid
39 mm, 15 mm
Price
1.030,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
Jars, made of PS, clear, with closing lid.
  • Good chemical resistance
  • Easy to clean
Suitable for ointments, powders, and granules.
Specifikationer
  • Udv.Ø:
    39 mm
  • Højde:
    15 mm
  • Seq:
    05