Storage bags
Storage bags, transparent, with ground weld., 0,075 mm, 260×400 mm
Katalog # SEMA2450
Leverandør: SEMADENI
Gå til:
Product Details & Documents
Storage bags in transparent LDPE, with bottom weld.
They are also suitable for liquids but without side weld.
Packaging: Minimum quantity:100 pieces
Specifikationer
- Beskrivelse:Storage bags, transparent, with ground weld.
- Dimensioner:260×400 mm
- Seq:07
- Tykkelse:0,075 mm