Clamps
15 mm, lyseblå
Katalog # SEMA1077
Leverandør: SEMADENI
Gå til:
Product Details & Documents
Clamps made of polyacetal (POM), in light blue.
- Chemical and temperature resistant
- Sticks (80 mm high) can be cut to the desired height
Specifikationer
- Spændvidde:15 mm
- Farve:lyseblå
- Seq:03