Clamps
Stick, POM, 80 mm high
Katalog # SEMA1084
Leverandør: SEMADENI
Gå til:
Product Details & Documents
Clamps made of polyacetal (POM), in light blue.
- Chemical and temperature resistant
- Sticks (80 mm high) can be cut to the desired height
Specifikationer
- Beskrivelse:Stick, POM, 80 mm high