Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Clamps

Clamps

45 mm, lyseblå
Katalog # SEMA1082
Leverandør:   SEMADENI
Clamps
Katalog # SEMA1082
Leverandør:  SEMADENI
Clamps
Clamps

45 mm, lyseblå

Price
23,20 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Clamps made of polyacetal (POM), in light blue.

  • Chemical and temperature resistant
  • Sticks (80 mm high) can be cut to the desired height
Specifikationer
  • Spændvidde:
    45 mm
  • Farve:
    lyseblå
  • Seq:
    08
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.