Orders LinkContactUsLinkComponent
Electrode Cables
Electrode cable, 2×2 mm, for connecting temperature sensors with Metrohm plug-in head G with 780, 781, 826, 827 pH/Ion meters and all Titrandos, 1 m
Katalog # METR6.2104.140
Leverandør:   METROHM
Electrode Cables
Katalog # METR6.2104.140
Leverandør:  METROHM
Electrode Cables
Electrode cable, 2×2 mm, for connecting temperature sensors with Metrohm plug-in head G with 780, 781, 826, 827 pH/Ion meters and all Titrandos, 1 m
Price
1.690,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Electrode cable, 2×2 mm, for connecting temperature sensors with Metrohm plug-in head G with 780, 781, 826, 827 pH/Ion meters and all Titrandos
  • Længde:
    1 m
  • Seq:
    09