Orders LinkContactUsLinkComponent
Skuffe racks til cryobokse
8 (2×4), 140×558×174 mm
Katalog # 479-1675
Leverandør:   TENAK
Skuffe racks til cryobokse
Katalog # 479-1675
Leverandør:  TENAK
Skuffe racks til cryobokse
Skuffe racks til cryobokse
8 (2×4), 140×558×174 mm
Price
1.420,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Rustfrit stål.
  • Racks til 5 forskellige højder æsker
  • Kan bruges i alle typer frysere
  • Passer til alle standard cryobokse
For upright freezers. These racks ensure a more ergonomic operating process. The cryoboxes are placed in drawers in the sliding shelf rack, increasing flexibility. Measure the inside height and depth of your freezer in order to choose the right rack.
Ordering information: Delivered with or without cryoboxes.
Specifikationer
  • Rum:
    8 (2×4)
  • B×D×H udv.:
    140×558×174 mm
  • Seq:
    64