Skuffe racks til cryobokse
6 (2×3), 140×421×174 mm
Katalog # 479-1673
Leverandør: TENAK
Gå til:
Product Details & Documents
Rustfrit stål.
- Racks til 5 forskellige højder æsker
- Kan bruges i alle typer frysere
- Passer til alle standard cryobokse
For upright freezers. These racks ensure a more ergonomic operating process. The cryoboxes are placed in drawers in the sliding shelf rack, increasing flexibility. Measure the inside height and depth of your freezer in order to choose the right rack.
Bestillingsoplysninger: Delivered with or without cryoboxes.
Documents
VWR for Cell Culture VWR® Lab bric-a-brac
Produktlitteratur
Specifikationer
- Rum:6 (2×3)
- B×D×H udv.:140×421×174 mm
- Seq:61