Skuffe racks til cryobokse
28 (7×4), 140×558×252 mm
479-1764
: TENAK
Rustfrit stål.
- Racks til 5 forskellige højder æsker
- Kan bruges i alle typer frysere
- Passer til alle standard cryobokse
For upright freezers. These racks ensure a more ergonomic operating process. The cryoboxes are placed in drawers in the sliding shelf rack, increasing flexibility. Measure the inside height and depth of your freezer in order to choose the right rack.
: Delivered with or without cryoboxes.
- Rum:28 (7×4)
- B×D×H udv.:140×558×252 mm
- Seq:08