HPLC syringes
Autosampler syringe, -, 250 µl, - mm, AS1000, AS3000
Katalog # 555-8225
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
These HPLC syringes are standard fixed needle syringes for Rheodyne/Valco™ injectors.
- Easy and accurate
- Reproducible manual injections
Applications include industrial chromatography.
Specifikationer
- Til:AS1000, AS3000
- Måler:-
- Volumen:250 µl
- Beskrivelse:Autosampler syringe
- Længde:- mm
- Seq:07