Thread adapters for pumps
Intrenal thread adapter, PP, brown, 71 mm - 50,8 mm BSP
Katalog # BURK0600-2002
Leverandør: BURKLE
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Intrenal thread adapter, PP, brown, 71 mm - 50,8 mm BSP
- Seq:07