Sample cups for analysers, ratiolab®
For Technicon T2 analysers, 3,5 ml
Katalog # RATI5700120
Leverandør: RATIOLAB
Gå til:
Product Details & Documents
Made of clear PS.
- Good chemical resistance
- Good temperature resistance
Analysis beakers offer the highest precision for use with robotic analytical systems.
Specifikationer
- Kapacitet:3,5 ml
- Beskrivelse:For Technicon T2 analysers
- Seq:04