Orders LinkContactUsLinkComponent
Pipette tips station, Corning® DeckWorks™
Pipette tips station, Corning® DeckWorks™
Rund spids, 0,1 - 10 µl, -, Transparent, 960 spidser/pakke
Katalog # CORN4141
   Reduktion af affald
Leverandør:   Corning
Pipette tips station, Corning® DeckWorks™
Katalog # CORN4141
Leverandør:  Corning
   Reduktion af affald
Pipette tips station, Corning® DeckWorks™
Pipette tips station, Corning® DeckWorks™
Rund spids, 0,1 - 10 µl, -, Transparent, 960 spidser/pakke
Price
3.190,00 kr
Log ind for tilgængelighed
1 * 4.800 Items
Product Details & Documents
These tip stations maximise bench space while minimising plastic waste. The stations contain 960 tips, available in extended length 10 µl and traditional 200 µl sizes.
  • Ideal single and multi-channel pipetting stations
  • Five-layer deck design (2×96) matches the microtiter plate format
  • Stable pipetting platform fits easily into sterile hood environments
  • Lids can be snap-positioned to expose a single side (96 tips) only
Applications include pre- and post-PCR procedures, cell culture and assays requiring large tip quantities. These tip stations are offered both sterile and non sterile.
Certified Free of: RNase, DNase, Pyrogens, PCR Inhibitors.
Both the hinged racks and reload decks within this system are manufactured with recycled plastic. The entire DeckWorks system is comprised of seven sizes of graduated pipette tips which are longer than standard tips to minimise the contamination issues often associated with traditional pipette tip styles. Sizes include 10, 20, 100, 200, 200, extended length, 300, and 1000 µl tips. The pipette tips are made in a controlled molding environment to ensure accuracy and precision across all tip volumes. Medical grade resins and production excellence allow for full certification as: nonpyrogenic, human DNA-free, RNase-/DNase-free, sterility assuarance level 10⁻³. All tips are manufactured from polypropylene.
Bestillingsoplysninger: Corning DeckWorks pipette tips are also available in bulk, hinged rack, barrier hinged rack, reload and reload starter kit configurations.
Documents
Specifikationer
  • Pakket:
    960 spidser/pakke
  • Steril:
    -
  • Volumen:
    0,1 - 10 µl
  • Spidstype:
    Rund spids
  • Spits farge:
    Transparent
  • Cat. No.:
    CORN4141
  • Environmentally Preferable:
    Reduktion af affald
Accessories
Produktinformation
  • Katalog #
    Til brug med
    Tilgængelighed
    Price
  • Pipettespidsracks, tomme, til 10/200/300 µl
    Pipettespidsracks, tomme, til 10/200/300 µl
    CORN4145
    Pipettespidsstation, DeckWorks™
    Log ind for tilgængelighed
    1 * 24 Items - 727,00 kr
Produktinformation
  • Katalog #
    Til brug med
    Tilgængelighed
    Price
  • Pipettespidsracks, tomme, med MultiGuard, til 1.000 µl
    Pipettespidsracks, tomme, med MultiGuard, til 1.000 µl
    CORN4146
    Pipettespidsstation, DeckWorks™
    Log ind for tilgængelighed
    1 * 16 Items - 666,00 kr