Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Mouse Recombinant PDGF AA (from E. coli)

Katalog # ABCAAB78590-10
Marketsource
Leverandør:   Abcam
Mouse Recombinant PDGF AA (from E. coli)
Katalog # ABCAAB78590-10
Leverandør:  Abcam
Marketsource
CAS nummer:  
Mouse Recombinant PDGF AA (from E. coli)
Mouse Recombinant PDGF AA (from E. coli)
CAS nummer:  
Price
3.260,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Recombinant mouse PDGF AA protein is a Mouse Full Length protein, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Konjugation:
    Unconjugated
  • Form:
    Lyophilised
  • Protein/peptide name:
    PDGF AA
  • Protein synonyms:
    Platelet-derived growth factor subunit A|PDGF subunit A|PDGF-1|Platelet-derived growth factor A chain|Platelet-derived growth factor alpha polypeptide|Pdgfa
  • Biologisk aktivitet:
    Standard
  • Dyrefri:
    N
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB78590-10
  • Protein/peptide type:
    Recombinant
  • Renhed:
    >95%
  • Tested applications:
    SDS-PAGE
  • Kilde:
    E. coli
  • Species:
    Mouse
  • Opbevaring betingelser:
    Store at –20 °C

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.