Depth filter capsules, Supracap™ 100
Product Details & Documents
Supracap™ 100 depth filter capsules with PP housing are scalable capsules with Seitz® high performance depth filter media. They are suitable for process development and pilot scale applications.
- Seamless linear scalability
- Completely disposable
- Encapsulated design reduces operator exposure to potential biohazards
Low hold-up volume allows for increased product recovery and requires low post-use rinse volumes.
Applications include biopharmaceuticals, mammalian cell cultures, therapeutic protein, vaccines, blood plasma proteins and serum, yeast, E.coli lysates and refolds.
The Seitz® Bio 20 media grade is made of highly purified natural and modified celluloses and is free from inorganic materials. Seitz® Bio 20 media is ideal for ion sensitive products such as therapeutic proteins, dialysis solutions, and protein-based diagnostics.
Seitz® P-series media are comprised of cellulose fibers, filter aids (diatomaceous earth and perlite), and resins. This grade is recommended for coarse filtration in the biotechnological and pharmaceutical industries.
Seitz® HP-series media are feature two distinct layers of Seitz® P-series depth filter sheets that feature a more permeable layer followed by a less permeable layer (PDD1, PDE2, PDH4 and PDK5). Seitz® HP-series media performs exceptionally well in low viability, high solids-containing applications, and with wider distribution of particle sizes in biotech applications.
: Manufactured under a quality management system certified to ISO 9000 and ISO 9001, and an environmental management system certified to ISO 14001.
Specifications
- Filtreringsområde:250 cm²
- Porestørrelse:0,5 - 15,0 µm
- Model:Seitz® PDH4 media with 1 - 1-1/2" sanitary flange connections, 154×213 mm
- Seq:13
- Cat. No.:PALLNP5LPDH41
- Max. temperature:40 °C
- Maks. tryk:3 bar
- Indløb/udløb:1 - 1-1/2" sanitær flange
- Ø×L:154×213 mm



