Precolumn filters, ColumnSaver
PEEK, 10-32, 2,0 µm
Katalog # 55215-U
Leverandør: Merck
Gå til:
Product Details & Documents
Precolumn filter kit has a standard 10-32 fitting and fits direct to most column types without introducing an additional dead volume.
Specifikationer
- Porøsitet:2,0 µm
- Gevind:10-32
- Materiale:PEEK
- Seq:02