Screw top vials
Vials, 40,00 ml, 28×95 mm, ravfarvet, No, 24-400
SIAL27382
: Merck
- Kasket inkluderet:No
- Cap størrelse:24-400
- Diameter×H:28×95 mm
- Kapacitet:40,00 ml
- Farve:ravfarvet
- Beskrivelse:Vials
- Seq:30
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
#: 20864.320
: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83640.320
: VWR Chemicals
Natriumhydroxid 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution
#: 31770.294
: VWR Chemicals
Vand, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83645.320
: VWR Chemicals