Orders LinkContactUsLinkComponent
Screw top vials
Vials, 40,00 ml, 28×95 mm, klar, No, 24-400
Katalog # SIAL27379
Leverandør:   Merck
Screw top vials
Katalog # SIAL27379
Leverandør:  Merck
Screw top vials
Vials, 40,00 ml, 28×95 mm, klar, No, 24-400
Price
1.480,00 kr
Log ind for tilgængelighed
1 * 1 Each
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Specifikationer
  • Kasket inkluderet:
    No
  • Cap størrelse:
    24-400
  • Diameter×H:
    28×95 mm
  • Kapacitet:
    40,00 ml
  • Farve:
    klar
  • Beskrivelse:
    Vials
  • Seq:
    19