Screw top vials
Vials, silanised, 4,00 ml, 15×45 mm, klar, No, 13-425
SIAL27114
: Merck
- Kasket inkluderet:No
- Cap størrelse:13-425
- Diameter×H:15×45 mm
- Kapacitet:4,00 ml
- Farve:klar
- Beskrivelse:Vials, silanised
- Seq:11