Orders LinkContactUsLinkComponent
Screw top vials
Screw top vials
Vials, 22,00 ml, 23×85 mm, ravfarvet, No, 20-400
Katalog # SIAL27073-U
Leverandør:   Merck
Screw top vials
Katalog # SIAL27073-U
Leverandør:  Merck
Screw top vials
Vials, 22,00 ml, 23×85 mm, ravfarvet, No, 20-400
Price
976,00 kr
Log ind for tilgængelighed
1 * 100 Items
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Specifikationer
  • Kasket inkluderet:
    No
  • Cap størrelse:
    20-400
  • Diameter×H:
    23×85 mm
  • Kapacitet:
    22,00 ml
  • Farve:
    ravfarvet
  • Beskrivelse:
    Vials
  • Seq:
    29