Orders LinkContactUsLinkComponent
OVS sampling tubes, ORBO™
OVS sampling tubes, ORBO™
ORBO™ 49P (OVS) Supelpak™-20 specially treated Amberlite® XAD®-2 (20/40), 270/140 mg RC,GFF,F,F (specially cleaned) separators, OSHA Versatile Sampler (OVS), O.D.×L: 13-8×75 mm, package of 10 ea
Catalog # SUPL20350
Supplier:   Merck
OVS sampling tubes, ORBO™
Catalog # SUPL20350
Supplier:  Merck
OVS sampling tubes, ORBO™
ORBO™ 49P (OVS) Supelpak™-20 specially treated Amberlite® XAD®-2 (20/40), 270/140 mg RC,GFF,F,F (specially cleaned) separators, OSHA Versatile Sampler (OVS), O.D.×L: 13-8×75 mm, package of 10 ea
Price
2.180,00 kr
Log in for availability
1 * 10 Items
Product Details & Documents
The OSHA Versatile Sampler (OVS) is a unique tube design with the adsorbent and filter combined in one tube for collection of both vapors and aerosols.
  • Packed with specially cleaned low background porous polymers
  • Most common sampling rate is 1 L/min to obtain a sample volume from 60 to 480 L
It′s most common use is for sampling pesticides but can also be used for other applications.
Specifications
  • Beskrivelse:
    ORBO™ 49P (OVS) Supelpak™-20 specially treated Amberlite® XAD®-2 (20/40), 270/140 mg RC,GFF,F,F (specially cleaned) separators, OSHA Versatile Sampler (OVS), O.D.×L: 13-8×75 mm, package of 10 ea
  • Seq:
    02