Accessories
Gelatin filters, wavelength 600 nm, bandpass ± 5 nm
Katalog # SHEW47189150
Leverandør: SHERWOOD SCIENTIFIC INSTRUMENT
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Gelatin filters, wavelength 600 nm, bandpass ± 5 nm
- Seq:03