Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Nitrile gloves, Venitactyl

Nitrile gloves, Venitactyl

Powdered, 6,5, blå, 240 mm, 100/box
Catalog # DELPV1420B1000/6.5
Supplier:   DELTA PLUS
Nitrile gloves, Venitactyl
Catalog # DELPV1420B1000/6.5
Supplier:  DELTA PLUS
Nitrile gloves, Venitactyl
Nitrile gloves, Venitactyl

Powdered, 6,5, blå, 240 mm, 100/box

Price
90,40 kr
Log in for availability
1 * 100 Items
Product Details & Documents

These disposable gloves provide protection against chemical products (occasional contact).

  • Non sterile, powdered or powder-free, smooth finish
  • Waterproof and airtight
  • Flexibility, elasticity and tactility
  • Latex-free: No allergies risks
  • Blue colour makes them easy to use: Ideal for use in the food industry

Suitable for use in food processing, industrial environments, chemical industries and in hygiene environments.

Certifications: EN 455-1/2-/3, EN ISO 374-1 Type B (JKT), EN ISO 374-5 VIRUS; AQL 1,5
Food contact approved

Specifications
  • Handske størrelse:
    6,5
  • Pakket:
    100/box
  • Farve:
    blå
  • Længde:
    240 mm
  • Seq:
    01
  • Model:
    Powdered
  • Cat. No.:
    DELPV1420B1000/6.5
  • Ambidextrous/Hånd-specifik:
    Ambidextrous
  • Godkendt til fødevarekontakt:
    +
  • Med pudder/pudderfri:
    Med pudder
  • Latex/latex-fri:
    Latex-fri
  • EN norm:
    EN 455-1/2-/3, EN ISO 374-1 Type B (JKT), EN ISO 374-5 VIRUS
  • Engangs/Genbrugelig:
    Engangs
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.