6-Methoxyindol
Glass bottle for solids, 25 g
Katalog # COBBIN-0054-25G
Marketsource
Leverandør: COMBI-BLOCKS
Gå til:
Product Details & Documents
Synonymer: 6-Methoxy-1H-indole, 1H-Indol-6-yl methyl ether, Indol-6-yl methyl ether
- CAS nummer:3189-13-7
- Formel:C₉H₉NO
- MDL nummer:MFCD00022780
- Molecular Weight:147.18 g/mol
- Opbevaringstemperatur:Fryser
- EINECS:221-689-6
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:25 g



