Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
6-Methoxyindol

6-Methoxyindol

Glass bottle for solids, 5 g
Katalog # COBBIN-0054-5G
Irriterende
Marketsource
Leverandør:   COMBI-BLOCKS
6-Methoxyindol
Katalog # COBBIN-0054-5G
Leverandør:  COMBI-BLOCKS
Marketsource
CAS nummer:  3189-13-7
6-Methoxyindol

Glass bottle for solids, 5 g

CAS nummer:  3189-13-7
Price
94,28 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
Synonymer:  6-Methoxy-1H-indole, 1H-Indol-6-yl methyl ether, Indol-6-yl methyl ether
  • CAS nummer:3189-13-7
  • Formel:C₉H₉NO
  • MDL nummer:MFCD00022780
  • Molecular Weight:147.18 g/mol
  • Opbevaringstemperatur:Fryser
  • EINECS:221-689-6
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.