Orders LinkContactUsLinkComponent
Instrument dishes
Instrument dishes
500 ml, 110×210×35 mm
Catalog # BURK4230-2050
Supplier:   BURKLE
Instrument dishes
Catalog # BURK4230-2050
Supplier:  BURKLE
Instrument dishes
Instrument dishes
500 ml, 110×210×35 mm
Price
165,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Trays for various uses. In laboratories, doctors’ surgeries, hospitals, pharmacies, the foodstuffs industry - wherever there is a need for clean storage of instruments, equipment, goods and precision parts.
  • Melamine, white
  • Tray without lid
Lid available as accessory.
Specifications
  • Kapacitet:
    500 ml
  • B×D×H:
    110×210×35 mm