PREGNANCY TEST CASETTE 25IU 1 * 10 Items
Katalog # SUREGHCGBC10
Leverandør: SURESCREEN
Frequently Bought Together
Natriumhydroxid ≥98%, pellets, GPR RECTAPUR®
Leverandør: VWR Chemicals
Ammoniakvand 25%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
Leverandør: Merck
Formaldehyd 4% stabilised, buffered (pH 7,0 ±0,1)
Leverandør: VWR Chemicals
Test strips for peracetic acid, MQuant®, Supelco®
Leverandør: Merck
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Leverandør: VWR Chemicals
Papir-pH-indikatorer, MQuant®, Supelco®
Leverandør: Merck
VWR® Rota®, pH indicator paper, 1 to 14
Leverandør: VWR Chemicals
Teststrips, MQuant™, Supelco®
Leverandør: Merck
Giemsa's farve opløsning til mikroskopi, Sigma-Aldrich®
Leverandør: Merck
Natriumhydroxid, piller, EMSURE® for analysis, Supelco®
Leverandør: Merck
Teststrimler, nitrat, MQuant™, Supelco®
Leverandør: Merck
External thread cryogenic vials
Leverandør: Corning
Natriumhypochlorit (3,5% Cl₂) in aqueous solution, GPR RECTAPUR®
Leverandør: VWR Chemicals
pH-teststrimler, MQuant®, Supelco®
Leverandør: Merck
Multiformat plates, silica gel 60, unmodified layers, glass backed
Leverandør: Merck
Test strips, peroxide, Dosatest
Leverandør: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Leverandør: VWR Chemicals
Cryogenic vials, CryoELITE®, WHEATON®
Leverandør: DWK Life Sciences