Orders LinkContactUsLinkComponent
TE Buffers 100X
TE Buffers 100X
TE buffer, 100X, 8,0, Plastic bottle, 250 mL
Katalog # 786-752
Leverandør:   G-Biosciences
TE Buffers 100X
Katalog # 786-752
Leverandør:  G-Biosciences
CAS nummer:  
TE Buffers 100X
TE buffer, 100X, 8,0, Plastic bottle, 250 mL
CAS nummer:  
Price
653,00 kr
Log ind for tilgængelighed
1 * 250 mL
Product Details & Documents
TE Buffer (Tris-EDTA Buffer) is a common molecular biology buffer that is used to protect DNA and RNA from degradation. The Tris buffering agent and EDTA metal chelating properties help protect DNA and RNA.
Specifikationer
Specifikationer
  • Pakning:
    Plastic bottle
  • Pakn:
    250 mL
  • Beskrivelse:
    TE buffer, 100X
  • pH-værdi:
    8,0