X-Gal
Product Details & Documents
5-Bromo-4-chloro-3-indolyl beta-D-galactopyranoside is most commonly known as X-Gal. X-gal is a histochemical substrate for beta-galactosidase which cleaves X-Gal to yield an insoluble blue precipitate.
Molecular Biology Grade
- Purity: Greater than 99% (HPLC)
- Solution: Clear (2% in DMF)
- Biological Activity: Blue colour with Lambda, pUC, and M13
X-Gal is a mainstay in molecular biology cloning applications in which it is used to detect the activity of beta-galactosidase. X-Gal is used to detect the insertion of foreign DNA into the lacZ region of a plasmid DNA. Insertion of DNA into the lacZ region results in the loss of beta-galactosidase activity. Bacteria cells that retain active beta-galactosidase will result in characteristic blue colonies. Successful disruption of the lacZα gene disrupts the α-complemtation of the beta-galactosidase gene and the precipitate does not form, resulting in white colonies.
Many other applications also use X-Gal as a substrate to detect beta-galactosidase activity. These include beta-galactosidase -antibody linked immunoassays and immunohistochemistry, coliphage detection based on beta-galactosidase induction and the detection of micrometastasis formation during tumor progession.
X-gal is commonly used in conjunction with IPTG for blue/white screening.
- CAS nummer:7240-90-6
- Formel:C₁₄H₁₅BrClNO₆
- MDL nummer:MFCD00005666
- Molecular Weight:408,63 g/mol
Specifikationer
Specifikationer
- Pakning:Vial
Specifikationer
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



