Ampicillin, natriumsalt
Glass bottle for solids, 5 g
A051-B
: D(-)-α-Aminobenzylpenicillin Sodium Salt, Ampicillin Na, Ampicillin sodium salt, D-(−)-α-Aminobenzylpenicillin sodium salt Ampicillin sodium salt
- :69-52-3
- :C₁₆H₁₈N₃NaO₄S
- :MFCD00064313
- :371.39 g/mol
- :Køleskab
- :200-708-1
- Pakning:Glass bottle for solids
- Pakn:5 g